Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Tri-Methyl K14]

Leverandør: Bioworld Technology
BS7525
BWRLBS7525EA 4900 NOK
BWRLBS7525
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Tri-Methyl K14]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, Tri methylated at K14.

Histone H3 (Tri-Methyl K14) pAb detects endogenous levels of histone H3 only when Tri-methylated at Lys14.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Tri-Methyl K14
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR