Anti-Histone H4 Rabbit Polyclonal Antibody [clone: Tri-Methyl K20]

Leverandør: Bioworld Technology
BS7676
BWRLBS7676EA 4900 NOK
BWRLBS7676
Anti-Histone H4 Rabbit Polyclonal Antibody [clone: Tri-Methyl K20]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H4, Tri methylated at K20.

Histone H4 (Tri-Methyl K20) pAb detects endogenous levels of histone H4 only when Tri-methylated at Lys20.

Type: Primary
Antigen: Histone H4
Clonality: Polyclonal
Clone: Tri-Methyl K20
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR