Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Leverandør: Avantor
C0001EA 925 NOK
C0001
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Fittings og kontakter
Avantor® ACE® PEEK one-piece column couplers are ideal for connecting analytical or semi-preparative columns to stand-alone guard cartridge holders. They can also be used to connect two or more HPLC columns in series or to make similar HPLC connections.
  • Universal connector, ultra-low dead volume
  • Pressure rated to 350 bar
  • Reusable, female-female connector
  • Inert and biocompatible