Avantor® ACE®, Preparative Column Coupler
Leverandør: Avantor
C0002EA 925 NOK
C0002
Avantor® ACE®, Preparative Column Coupler
Fittings og kontakter
Column coupler made from stainless steel/PEEK suitable for connecting preparative columns to stand-alone guard cartridge holder, or for making similar HPLC connections.
Ideally suited to connect 30 mm id preparative columns to H0009 cartridge holder.