6-Carboxyfluorescein, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

C0662-50MG
C0662-50MGEA 1660 NOK
C0662-50MG
6-Carboxyfluorescein, Sigma-Aldrich®
6-Carboxyfluorescein

Fluorescent dye.

Formel: C₂₁H₁₂O₇
MW: 376.32 g/mol
Smeltepunkt: >300 °C (lit.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036873
CAS-nummer: 3301-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9983c95b11ec00cf9b0fc86b5f13cef79b7675e&vid=5eef1ef61ae9547208bbc0603f984ca16922e43f&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR