Coproporphyrin I tetramethyl ester ≥90% (by HPLC), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

C4529-1MG
C4529-1MGEA 1450 NOK
C4529-1MG
Coproporphyrin I tetramethyl ester ≥90% (by HPLC), Sigma-Aldrich®
Coproporphyrin I tetramethyl ester

Coproporphyrin I tetramethyl ester. ≥90% (HPLC) (Tetramethyl 3,8,13,18-tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropionate)

Smeltepunkt: 252…254 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00079038
CAS-nummer: 25767-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17b7b45cf9d3a0db21aea59f8d46fe60a14c8e8b&vid=b0c5ba33be7595bb7bc79f21292897dd8a3837ed&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR