Carmine, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: CI 75470

M-Clarity™ quality level = MQ200

C6152-25G
C6152-25GEA 4010 NOK
C6152-25G
Carmine, Sigma-Aldrich®
Carmine
Formel: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
MDL nummer: MFCD00167028
CAS-nummer: 1390-65-4
EINECS: 215-724-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR