Kongorødt, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger


M-Clarity™ quality level = MQ200

C6767-100G C6767-25G
C6767-100GEA 1020 NOK
C6767-100G C6767-25G
Kongorødt, Sigma-Aldrich®
Kongorødt
Formel: C₃₂H₂₂N₆Na₂O₆S₂
Smeltepunkt: > 360 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004028
CAS-nummer: 573-58-0
EINECS: 209-358-4
Merck Indeks: 12,02562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b99da120fc58f7f664a30e809aff2930cdb0d363&vid=818696761bbb066454f93ef8d9bc89285322112d&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR