Refractive Index Liquids, Series M
Leverandør: CARGILLE
1815Y1.750
CARG1815Y1.750EA 12075 NOK
CARG1815Y1.750
Refractive Index Liquids, Series M
Standarder Brytningsindeks standarder
Refractive index liquids have a broad range of applications in diversified fields.
  • Light yellow liquid
  • Slight unpleasant odour
Adjustment: ±0,0005
Choose from range nD 1,705 to 1,800