N-Arachidonoyl-L-Serine 1% in ethanol , 98%

Leverandør: Cayman Chemical

10005455-10
CAYM10005455-10EA 2350 NOK
CAYM10005455-10
N-Arachidonoyl-L-Serine 1% in ethanol , 98%
N-Arachidonoyl-L-Serine

Arachidonoyl amides of both amino acids and neurotransmitters such as dopamine have been previously reported in the literature.
(1)N-Arachidonoyl-L-serine (ARA-S) is one such recently isolated endocannabinoid with an unusual activity profile. ARA-S does not bind to central cannabinoid (CB1) and peripheral cannabinoid (CB2) receptors or vanilloid receptor 1 (VR1). Like cannabidiol, ARA-S (5 mg/kg) antagonizes the hypotensive effects of a 10 mg/kg IV bolus of abnormal cannabidiol (Abn-CBD) in an anesthetized rat blood pressure model.
(2)However, similar to Abn-CBD, ARA-S relaxes isolated rat mesenteric arteries and abdominal aorta as well as increases phosphorylation of Akt and mitogen-activated protein kinase (MAPK) in HUVEC.
(3)The precise mechanisms of action by ARA-S and Abn-DBD in various vascular preparations appears to be different and requires further investigation.


  • Purity: ≥98%
  • Formulation: A solution in ethanol
  • Storage: -20°C
  • Stability: ≥ 1 year


Product Description References
1. Bisogno, T.,Melck, D.,Bobrov, M.Y., et al. N-acyl-dopamines: Novel synthetic CB1 cannabinoid-receptor ligands and inhibitors of anandamide inactivation with cannabimimetic activity in vitro and in vivo. Biochemistry Journal 351, 817-824 (2001).
2. Milman, G.,Maor, Y.,Horowitz, M., et al. Arachidonoyl-serine, an endocannabinoid-like bioactive constituent of rat brain. 14th Annual Symposium on the Cannabinoids 133-133 (2004).
3. Milman, G.,Maor, Y.,Abu-Lafi, S., et al. N-arachidonoyl L-serine, an endocannabinoid-like brain constituent with vasodilatory properties. Proceedings of the National Academy of Sciences of the United States of America 103(7), 2428-2433 (2006).


Warning - this product is not for human or veterinary use.

Formel: C₂₃H₃₇NO₄
MDL nummer: MFCD08062151
CAS-nummer: 187224-29-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR