Cerulenin ≥98%, crystalline solid

Leverandør: Cayman Chemical
Warning

10005647-10 10005647-5 10005647-50
CAYM10005647-10EA 6525 NOK
CAYM10005647-10 CAYM10005647-5 CAYM10005647-50
Cerulenin ≥98%, crystalline solid
Cerulenin

A number of tumor cells and cell lines have been observed to have highly upregulated expression and activity of fatty acid synthase (FAS). Inhibition of FAS by the irreversible inhibitor cerulenin leads to cytotoxicity and apoptosis in human cancer cell lines, an effect believed to be mediated by the accumulation of malonyl-coenzyme A in cells with an upregulated FAS pathway. Cerulenin also leads to profound weight loss and feeding inhibition in both high-fat diet wild type obese and leptin-deficient ob/ob mice.

Formel: C₁₂H₁₇NO₃
MW: 223.272 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00077686
CAS-nummer: 17397-89-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR