Human recombinant Renin
Leverandør: Cayman Chemical
10006217-25
CAYM10006217-25EA 12050 NOK
CAYM10006217-25
Human recombinant Renin
Proteiner
Renin is an aspartyl protease of approximately 40 kDa produced in the kidney by juxtaglomerular cells of the macula densa.
  • Renin converts angiotensinogen into angiotensin I
  • Mature and active
  • 340 amino acid protease
Native human renin is a glycoprotein derived from preprorenin by sequential cleavage of 20 and 46 amino acid residues from the N-terminus of the 406 amino acid precursor.