Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Cayman Chemical
10006588-1
CAYM10006588-1EA 2950 NOK
CAYM10006588-1
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR