SREBP-2 transcription factor assay kit
Leverandør: Cayman Chemical
10007819-96
CAYM10007819-96EA 15475 NOK
CAYM10007819-96
SREBP-2 transcription factor assay kit
Assay Cellulær analyse
96 well assay for measurement of SREBP-2.
  • A sensitive, non-radioactive method of detecting SREBP-2 from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
  • Nuclear extraction kit is available to aid in the isolation of nuclear and cytoplasmic fractions from cell lysates or tissue homogenates
Lipid homeostasis in vertebrate cells is regulated by a family of transcription factors called sterol regulatory element-binding proteins (SREBP’s). SREBP’s directly activate the expression of over 30 genes involved in the synthesis and uptake of cholesterol, fatty acids, triglycerides, and phospholipids. Three different SREBP isoforms designated SREBP-1a, SREBP-1c, and SREBP-2 are encoded by two different genes and belong to the basic helix-loop-helix-leucine zipper (bHLH-ZIP) family of transcription factors. SREBP-2 performs a critical role in the transcriptional regulation of genes involved in cholesterol synthesis and uptake including HMG-CoA synthase, HMG-CoA reductase, and the LDL receptor. Cayman’s SREBP-2 Transcription Factor Assay is a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates. A 96 well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA). A specific double stranded DNA (dsDNA) sequence containing the SREBP response element is immobilized to the wells of a 96 well plate. SREBP contained in a nuclear extract, binds specifically to the SREBP response element. SREBP is detected by addition of specific primary antibody directed against SREBP. A secondary antibody conjugated to HRP is added to provide a sensitive colorimetric readout at 450 nm.
Leveringsinformasjon: Kit contains assay buffer, reagent A, positive control, binding buffer, primary antibody, wash buffer concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover sheet, developing and stop solution.