cis-11-Methyl-2-dodecenoic acid

Leverandør: Cayman Chemical

Synonymer: (Z)-11-Methyldodec-2-enoic acid, DSF, Diffusible signal factor

10008123-1 10008123-10 10008123-5
CAYM10008123-1EA 2575 NOK
CAYM10008123-1 CAYM10008123-10 CAYM10008123-5
cis-11-Methyl-2-dodecenoic acid
cis-11-Methyl-2-dodecenoic acid
Formel: C₁₃H₂₄O₂
CAS-nummer: 677354-23-3
FN: 1231
ADR: 3,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR