2-Tiobarbitursyre

Leverandør: Cayman Chemical

10009199-1
CAYM10009199-1EA 2250 NOK
CAYM10009199-1
2-Tiobarbitursyre
2-Tiobarbitursyre
Formel: C₄H₄N₂O₂S
Kokepunkt: 419 °C (1013 hPa)
Smeltepunkt: 235 °C
Tetthet: 1,661 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006674
CAS-nummer: 504-17-6
EINECS: 207-985-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR