Cholesterol cell-based detection assay kit
Leverandør: Cayman Chemical
10009779-192
CAYM10009779-192EA 8850 NOK
CAYM10009779-192
Cholesterol cell-based detection assay kit
Assay Cellulær analyse
Cholesterol Cell-Based Detection Assay Kit is a 192-well assay for measurement of cholesterol.
  • Detect cholesterol uptake and localisation within cultured cells
  • Employs filipin III, a probe that fluoresces in the presence of cholesterol
  • An inhibitor of cholesterol trafficking is included as a positive control
The mechanism for the movement of cholesterol from intracellular sites to their ultimate cellular destination is an unresolved question of fundamental importance to cell biology and medicine. Thus, defining mechanisms of intracellular cholesterol transport and identifying the cellular factors involved are therefore of great interest. The Cholesterol Cell-Based Detection Assay Kit provides a simple fluorometric method to study mechanisms and biological factors that regulate cholesterol metabolism or movement within cells.
Leveringsinformasjon: The kit includes filipin III, fixative, and wash buffer in a ready to use format.