Anti-COX-2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Cayman Chemical
10010096-1
CAYM10010096-1EA 11050 NOK
CAYM10010096-1
Anti-COX-2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR