S-Glutathionylated Protein Detection Kit
Leverandør: Cayman Chemical
10010721-1
CAYM10010721-1EA 10925 NOK
CAYM10010721-1
S-Glutathionylated Protein Detection Kit
Assay Cellulær analyse
A convenient method for the direct visualisation of S-glutathionylated proteins in whole (permeabilised) cells.
  • Direct visualisation of S-glutathionylated proteins in whole (permeabilised) cells by flow cytometry and microscopy
  • Visualisation using colorimetric or fluorescence detection
  • Reagents provided to test up to 30 samples
Glutathione (GSH) is involved in maintenance of protein sulfhydryl reduction status. PSSG may modify the activity of a large number of cell proteins, including metabolic, structural, cytoskeletal, and signaling proteins. Cayman’s PSSG provides a convenient method for the direct visualisation of S-glutathionylated proteins in whole (permeabilised) cells by flow cytometry and microscopy as well as avidin overlay analysis. This cell-based assay starts with the modification of protein free-thiols groups followed by enzymatic cleavage of any PSSG adducts present in the sample. Biotinylation of the newly-formed protein free-thiols provides the basis for visualisation using streptavidin-based colorimetric or fluorescence detection. Reagents are provided to test three sets of 10 samples (most convenient) or up to thirty samples total at once if desired.
Bestill nå
SPESIFIKASJONER
Merkenavn CAYM