Okadaic acid

Leverandør: Cayman Chemical

10011490-25 10011490-50 10011490-100
CAYM10011490-25EA 1800 NOK
CAYM10011490-25 CAYM10011490-50 CAYM10011490-100
Okadaic acid
Okadaic acid
Formel: C₄₄H₆₈O₁₃
MDL nummer: MFCD00083455
CAS-nummer: 78111-17-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR