Anti-IgA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Cayman Chemical
10196-500
CAYM10196-500EA 3925 NOK
CAYM10196-500
Anti-IgA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Primary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IgA
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Mouse
Vert Rabbit
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Immunogen mouse IgA
Rensing purified by Protein A chromatography
Kyssabsorpsjon No
Lagringsbuffer PBS, pH 7,2, containing 50% glycerol, 0,1% BSA, and 0,02% sodium azide
Lagringstemp +4 °C
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR