Anti-Phosphothreonine Mouse Monoclonal Antibody [clone: 18F6]

Leverandør: Cayman Chemical
10221-100
CAYM10221-100EA 16150 NOK
CAYM10221-100
Anti-Phosphothreonine Mouse Monoclonal Antibody [clone: 18F6]
Antistoffer
Type: Primary
Antigen: Phosphothreonine
Clonality: Monoclonal
Clone: 18F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol pT
Antigen-navn Phosphothreonine
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 18F6
Reaktivitet
Vert Mouse
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
Format Ascites
Kyssabsorpsjon No
Lagringsbuffer containing 0,1% sodium azide
Lagringstemp –20 °C
Frakttemp. –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR