Anti-PDCD4 Rabbit Polyclonal Antibody

Leverandør: Cayman Chemical
10250-500
CAYM10250-500EA 9900 NOK
CAYM10250-500
Anti-PDCD4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PDCD4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR