Prostaglandin Screening Library III

Leverandør: Cayman Chemical

10503-50
CAYM10503-50EA 36700 NOK
CAYM10503-50
Prostaglandin Screening Library III
Assay Cellulær analyse

For A and J series prostaglandin analog screening.


  • Contains >75 compounds
  • Includes the ''A- and J-series'' prostaglandins


The Prostaglandin Screening Library III contains more than 75 ''A and J-series'' of prostaglandins in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The library may also include thromboxanes, lactones, stable PGH, prostacyclin mimic, metabolites, and potent PGA and PGJ analogs. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR