4-Methylthiobenzamide

Leverandør: Cayman Chemical

13229-250 13229-500 13229-1 13229-5
CAYM13229-250EA 545 NOK
CAYM13229-250 CAYM13229-500 CAYM13229-1 CAYM13229-5
4-Methylthiobenzamide
4-Methylthiobenzamide
Formel: C₈H₉NS
MDL nummer: MFCD00173750
CAS-nummer: 2362-62-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR