Anti-PARP Mouse Monoclonal Antibody [clone: N/A]

Leverandør: Cayman Chemical
13557-1
CAYM13557-1EA 14650 NOK
CAYM13557-1
Anti-PARP Mouse Monoclonal Antibody [clone: N/A]
Antistoffer
Type: Primary
Antigen: PARP
Clonality: Monoclonal
Clone: N/A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol PARP
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Mouse
Isotype IgG2b kappa
Western blot Yes
Flowcytometri Yes
Immunogen synthetic peptide containing amino acids near the 214/215 cleavage site of human PARP
Molekylvekt 116 kDa
Rensing purified by Protein G chromatography
Kyssabsorpsjon No
Lagringsbuffer PBS containing 0,05% BSA and 0,05% sodium azide
Lagringstemp –20 °C
Frakttemp. –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR