Anti-Cav1.2 Mouse Monoclonal Antibody [clone: S57-46]

Leverandør: Cayman Chemical
13702-100
CAYM13702-100EA 16150 NOK
CAYM13702-100
Anti-Cav1.2 Mouse Monoclonal Antibody [clone: S57-46]
Antistoffer
Type: Primary
Antigen: Cav1.2
Clonality: Monoclonal
Clone: S57-46
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol Cav1.2
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon S57-46
Reaktivitet Human,Rat,Mouse
Vert Mouse
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Immunogen fusion protein amino acids 1,507-1,733
Aminosyretall 1507 to 1733
Rensing purified by Protein G chromatography
Kyssabsorpsjon No
Lagringsbuffer PBS, pH 7,4, containing 50% glycerol and 0,09% sodium azide
Lagringstemp –20 °C
Frakttemp. –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR