Anti-NF kappaB (p65) Mouse Monoclonal Antibody (Biotin) [clone: 112A1021]

Leverandør: Cayman Chemical
13756-1
CAYM13756-1EA 13625 NOK
CAYM13756-1
Anti-NF kappaB (p65) Mouse Monoclonal Antibody (Biotin) [clone: 112A1021]
Antistoffer
Type: Primary
Antigen: NF kappaB (p65)
Clonality: Monoclonal
Clone: 112A1021
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR