Anti-MeCP2 Rabbit Polyclonal Antibody

Leverandør: Cayman Chemical
13775-1
CAYM13775-1EA 12050 NOK
CAYM13775-1
Anti-MeCP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MeCP2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR