TAK-242 ≥98%

Leverandør: Cayman Chemical

Synonymer: Resatorvid, (R)-Ethyl 6-(N-(2-chloro-4-fluorophenyl)sulphamoyl)cyclohex-1-enecarboxylate, 6R-[[(2-chloro-4-fluorophenyl)amino]sulphonyl]-1-cyclohexene-1-carboxylic acid, ethyl ester

13871-1 13871-10 13871-25 13871-5
CAYM13871-1EA 2300 NOK
CAYM13871-1 CAYM13871-10 CAYM13871-25 CAYM13871-5
TAK-242 ≥98%
TAK-242

Toll-like receptor 4 (TLR4) is activated by lipopolysaccharide (LPS), with LPS-binding protein, to initiate an innate immune response. Alternatively, hypoxia or ischemia/reperfusion can trigger signaling through TLR4. TAK-242 is a cell-permeable inhibitor of TLR4 signaling, blocking LPS-induced production of NO, TNF-α, IL-6, and IL-1β in macrophages with IC50 values of 1-11 nM. It does not affect signaling through other TLRs.3,4,5 TAK-242 is effective in vivo, suppressing the production of NO, TNF-α, IL-6, and IL-1β in mice treated with LPS when given intravenously.3 Mechanistically, TAK-242 acts by selectively binding TLR4 and preventing its association with adaptor proteins that are essential for signal transduction. TAK-242 is used to study signaling through TLR4 in a variety of pathologies, including sepsis and ischemia/reperfusion injury.

Formel: C₁₅H₁₇NO₄ClFS
MW: 361.8 g/mol
MDL nummer: MFCD11046362
CAS-nummer: 243984-11-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR