Puromycin dihydrochloride

Leverandør: Cayman Chemical
Warning

Synonymer: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

13884-25 13884-50 13884-100 13884-500
CAYM13884-25EA 1125 NOK
CAYM13884-25 CAYM13884-50 CAYM13884-100 CAYM13884-500
Puromycin dihydrochloride
Puromycin dihydrochloride
Formel: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00012691
CAS-nummer: 58-58-2
EINECS: 200-387-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR