Anti-I kappaB-zeta Rabbit Polyclonal Antibody

Leverandør: Cayman Chemical
13926-1
CAYM13926-1EA 12700 NOK
CAYM13926-1
Anti-I kappaB-zeta Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: I kappaB-zeta
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR