Anti-KLH Rabbit Polyclonal Antibody

Leverandør: Cayman Chemical
14046-1
CAYM14046-1EA 9900 NOK
CAYM14046-1
Anti-KLH Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR