Blasticidin S hydrochloride ≥95%, crystalline solid

Leverandør: Cayman Chemical
Danger

14499-10 14499-100 14499-25 14499-50
CAYM14499-10EA 1275 NOK
CAYM14499-10 CAYM14499-100 CAYM14499-25 CAYM14499-50
Blasticidin S hydrochloride ≥95%, crystalline solid
Blasticidin S hydrochloride

Blasticidin S is a peptidyl nucleoside antibiotic. It was first isolated from S. griseochromogenes. The structure of the pentahydrate form is determined using diffractometer data and, similar to the antibiotic amicetin, the hydrophilic and hydrophobic groups lie on either side of the molecule.


  • Created using total synthesis
  • Stability: ≥2 years


Used in research to select mammalian cells transfected with the blasticidin resistance gene bsr.


NB: Not for human or veterinary use.

Formel: C₁₇H₂₇ClN₈O₅
Lagringstemperatur: Fryseskap
MDL nummer: MFCD02091640
CAS-nummer: 3513-03-9
FN: 2811
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Purity ≥95%
λmax 269 nm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR