6-Cyano-7-nitroquinoxaline-2,3-dione ≥98%

Leverandør: Cayman Chemical

14618-5 14618-10 14618-25 14618-50
CAYM14618-5EA 1600 NOK
CAYM14618-5 CAYM14618-10 CAYM14618-25 CAYM14618-50
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98%
6-Cyano-7-nitroquinoxaline-2,3-dione

CNQX is a competitive, non-NMDA glutamate receptor antagonist (IC50s = 0.3 and 1.5 μM for AMPA and kainate receptors, respectively, versus IC50 = 25 μM for NMDA receptors).
This compound has been used to specifically target AMPA and kainate receptor responses and thus differentiate from that of NMDA receptors.

Formel: C₉H₄N₄O₄
MDL nummer: MFCD00069232
CAS-nummer: 115066-14-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR