7-Ethyl-10-hydroxycamptothecin ≥98%

Leverandør: Cayman Chemical
Danger

15632-10 15632-50 15632-5
CAYM15632-10EA 2900 NOK
CAYM15632-10 CAYM15632-50 CAYM15632-5
7-Ethyl-10-hydroxycamptothecin ≥98%
7-Ethyl-10-hydroxycamptothecin

SN-38 is the active metabolite of irinotecan, a derivative of the alkaloid camptothecin and acts as a potent inhibitor of DNA topoisomerase I. SN-38 is a more potent topopisomerase I inhibitor and more cytotoxic to HT-29 colon cancer cells (IC50 = 8.8 nM) compared to irinotecan (IC50 >100 nM).


  • SN-38 is in a form of crystalline solid
  • λmax at 224, 267, 331, 368, 383 nm

Formel: C₂₂H₂₀N₂O₅
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00871873
CAS-nummer: 86639-52-3
FN: 2811
ADR: 6.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR