Anti-dsDNA Mouse Monoclonal Antibody [clone: 2C4]

Leverandør: Cayman Chemical
15635-1
CAYM15635-1EA 9900 NOK
CAYM15635-1
Anti-dsDNA Mouse Monoclonal Antibody [clone: 2C4]
Antistoffer
Type: Primary
Antigen: dsDNA
Clonality: Monoclonal
Clone: 2C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR