Taurodeoxycholic acid sodium salt ≥95%

Leverandør: Cayman Chemical
Warning

Synonymer: Sodium 2-((R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulphonate, Taurodeoxycholate sodium salt, Sodium taurodeoxylate, Cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulphonate

15935-25 15935-5 15935-1
CAYM15935-25EA 13375 NOK
CAYM15935-25 CAYM15935-5 CAYM15935-1
Taurodeoxycholic acid sodium salt ≥95%
Taurodeoxycholic acid sodium salt

Taurodeoxycholic acid is a taurine-conjugated form of the secondary bile acid deoxycholic acid. Taurodeoxycholic acid stimulates chloride ion secretion through calcium-activated chloride ion channels and cystic fibrosis transmembrane conductance regulator (CFTR) in Calu-3 airway epithelial cell monolayers when applied basolaterally. Serum levels of taurodeoxycholic acid increase approximately 5-fold in within two hours during an oral lipid tolerance test in humans.

Formel: C₂₆H₄₄NO₆S·Na
MDL nummer: MFCD00003671
CAS-nummer: 1180-95-6

Bestill nå

Spesifikasjon Testresultater

Purity ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR