Imipenem monohydrate

Leverandør: Cayman Chemical

16039-10 16039-25 16039-100 16039-5
CAYM16039-10EA 2475 NOK
CAYM16039-10 CAYM16039-25 CAYM16039-100 CAYM16039-5
Imipenem monohydrate
Imipenem monohydrate

Imipenem is an intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0,5 - 2 µg/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species. Imipenem acts by interfering with bacterial cell wall synthesis.


  • A crystalline solid

Formel: C₁₂H₁₇N₃O₄S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD09753321
CAS-nummer: 74431-23-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR