Anti-AIF Rabbit Polyclonal Antibody

Leverandør: Cayman Chemical
160773-500
CAYM160773-500EA 9900 NOK
CAYM160773-500
Anti-AIF Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AIF
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR