(−)-Apomorphine hydrochloride hydrate ≥95%

Leverandør: Cayman Chemical
Danger

Synonymer: 5,6,6aR,7-tetrahydro-6-methyl-4h-dibenzo[de,g]quinoline-10,11-diol monohydrochloride hydrate

16094-10 16094-100 16094-50
CAYM16094-10EA 1025 NOK
CAYM16094-10 CAYM16094-100 CAYM16094-50
(−)-Apomorphine hydrochloride hydrate ≥95%
(−)-Apomorphine hydrochloride hydrate

(−)-Apomorphine is a classic dopamine receptor agonist that exhibits pKi values of 6,43, 7,08, 7,59, 8,36, and 7,83 for human recombinant D₁, D₂L, D₃, D₄, and D₅ receptors respectively. It produces biphasic effects on locomotor activity and displays anti-parkinsonian and neuroprotective actions.


NB: This product is not for human or veterinary use.

Formel: C₁₇H₁₇NO₂·HCl·xH₂O
Lagringstemperatur: Fryseskap
CAS-nummer: 58117-94-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR