Anti-DYKDDDDK Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Leverandør: Cayman Chemical
162150-1
CAYM162150-1EA 9900 NOK
CAYM162150-1
Anti-DYKDDDDK Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Antistoffer
Type: Tag
Antigen: DYKDDDDK
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR