Malonyl coenzyme A lithium salt ≥90%

Leverandør: Cayman Chemical

16455-1 16455-10 16455-5 16455-25
CAYM16455-1EA 1400 NOK
CAYM16455-1 CAYM16455-10 CAYM16455-5 CAYM16455-25
Malonyl coenzyme A lithium salt ≥90%
Malonyl coenzyme A lithium salt

Malonyl-CoA is formed by acetyl-CoA carboxylase-mediated carboxylation of acetyl-CoA. Fatty acid synthase catalyzes the NADPH-dependent condensation of malonyl-CoA and acetyl-CoA to produce palmitate. Proliferating human cancer cells upregulate this fatty acid synthesis pathway as a strategy for survival. High levels of malonyl-CoA, achieved through fatty acid synthase inhibition, proposed to contribute to cancer cell apoptosis.


Malonyl coenzyme A is a coenzyme A derivative is used in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane and also used as the extender unit in the synthesis of bacterial aromatic polyketides.

Formel: C₂₄H₃₇N₇O₁₉P₃SLi
MW: 859.6 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00057769
CAS-nummer: 108347-84-8

Bestill nå

Spesifikasjon Testresultater

Purity ≥90%
λmax 257 nm
Formulation A Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR