Biotin hydrazide ≥95%

Leverandør: Cayman Chemical

Synonymer: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide

18418-1 18418-100 18418-250 18418-500
CAYM18418-1EA 10375 NOK
CAYM18418-1 CAYM18418-100 CAYM18418-250 CAYM18418-500
Biotin hydrazide ≥95%
Biotin hydrazide

Biotin hydrazide is a biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups.


Used as a probe for determining protein carbonylation, an irreversible posttranslational modification resulting from the actions of reactive oxygen species or lipid oxidation products. The reaction of this probe is direct and does not require catalysts or reducing agents.

Formel: C₁₀H₁₈N₄O₂S
MDL nummer: MFCD00078532
CAS-nummer: 66640-86-6

Bestill nå

Spesifikasjon Testresultater

Purity ≥95%
Formulation A Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR