Anti-TBK1 mouse monoclonal antibody [clone: 4E6]

Leverandør: Cayman Chemical
25924-100
CAYM25924-100EA 11425 NOK
CAYM25924-100
Anti-TBK1 mouse monoclonal antibody [clone: 4E6]
Antistoffer
Type: Primary
Antigen: TBK1
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR