ZZW-115 trihydrochloride ≥95%

Leverandør: Cayman Chemical

Synonymer: N,N-Dimethyl-2-(4-(3-(2-(trifluoromethyl)-10H-phenothiazin-10-yl)propyl)piperazin-1-yl)ethanamine trihydrochloride, ZZW115 trihydrochloride, 10-[3-[4-[2-(Dimethylamino)ethyl]-1-piperazinyl]propyl]-2-(trifluoromethyl)phenothiazine trihydrochloride, ZZW 115 trihydrochloride, ZZW-115 hydrochloride

34974-1
CAYM34974-1EA 1750 NOK
CAYM34974-1
ZZW-115 trihydrochloride ≥95%
ZZW-115 trihydrochloride
Formel: C₂₄H₃₁F₃N₄S·3HCl
CAS-nummer: 10122-45-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR