Tween® 20 (Polysorbate)

Leverandør: Cayman Chemical

Synonymer: Polyoxyethylene sorbitan monolaurate

400035-1
CAYM400035-1EA 380 NOK
CAYM400035-1
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Formel: C₅₈H₁₁₄O₂₆
Tetthet: 1,1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00165986
CAS-nummer: 9005-64-5
EINECS: 500-018-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR