Nitrotyrosine IP Kit
Leverandør: Cayman Chemical
601220-1
CAYM601220-1EA 14350 NOK
CAYM601220-1
Nitrotyrosine IP Kit
Assay Cellulær analyse
For the capture and pulldown of nitrated proteins from cell and tissue lysates.
  • Determine change in tyrosine nitration in treated versus non-treated samples
  • Demonstrate target proteins are nitrated in vitro
  • Identify and characterise nitrated proteins by Western blot and proteomic analysis
Cayman’s Nitrotyrosine IP Kit allows for the capture and pulldown of nitrated proteins from cell lysates using a sorbent coupled with a nitrotyrosine monoclonal antibody. The nitrated proteins can be eluted from the resin, loaded to an SDS-PAGE gel, and probed using the included Nitrotyrosine Polyclonal Antibody or a user supplied target specific antibody. A positive control sample assures that the assay is working.