Anti-Ovalbumin IgG1 (Mouse) ELISA Kit
Leverandør: Cayman Chemical
700260-96
CAYM700260-96EA 9075 NOK
CAYM700260-96
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit
Assay ELISAs
For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.
  • Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
  • Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
  • Assay Range: 1.56 – 200 ng/ml with an LLOQ of 1.56 ng/ml
  • Rapid assay; get results in under 4 hours
  • LLOD: 0.006 U/ml
Cayman's Ovalbumin IgG1 (mouse) ELISA Kit is an immunometric (i.e.,'sandwich') assay which can be used to measure anti-ovalbumin of the IgG1 isotype in mouse plasma and serum without prior sample purification.