Uric acid assay kit
Leverandør: Cayman Chemical
700320-96
CAYM700320-96EA 5600 NOK
CAYM700320-96
Uric acid assay kit
Assay Cellulær analyse
Uric Acid Assay Kit is used to measure uric acid in serum, plasma and urine.
  • Assay 40 samples in duplicate
  • Assay range: 0,5 to 10 μM
  • Plate-based fluorometric measurement (ex 530 to 540 nm, em 585 to 595 nm)
Uric acid (urate) is the end product of human purine metabolism and is mainly excreted in urine. Many factors, including genetic components and acquired factors, such as obesity and alcohol consumption, influence serum uric acid concentrations. Hyperuricemia induces or facilitates gout, kidney stones, metabolic syndrome, hypertension, and renal and cardiovascular disease, while exercise-induced acute renal failure is a significant complication of renal hypouricemia.